
Toluene-4-sulphonamide ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:70-55-3
:
Formula:C₇H₉NO₂S
MDL Number:MFCD00011692
Boiling Pt:214°C
Melting Pt:136…138 °C
:171.22 g/mol
Storage Temperature:Ambient
EINECS:200-741-1
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:136.0 to 140.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together