
α,α,α-Trifluoro-p-toluoyl chloride ≥97.0% (by GC)
TCIAT1301-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
:
:208.57 g/mol
:68°C
:–3 °C
:1,4035 g/cm³ (20 °C)
:78°C
:Ambient
:206-342-9
:3265
:8,II
:GHS05
:329-15-7
:C₈H₄ClF₃O
:MFCD00000694