
α,α,α-Trifluoro-m-toluoyl chloride ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2251-65-2
:
Formula:C₈H₄ClF₃O
MDL Number:MFCD00000680
:208.57 g/mol
Boiling Pt:85°C
Density:1,4078 g/cm³ (20 °C)
EINECS:218-844-5
UN:3265
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together