
α,α,α-Trifluoro-o-toluoyl chloride ≥98.0% (by GC, titration analysis)
Catalog # TCIAT1280-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
About this item
:
:208.57 g/mol
Boiling Pt:90°C
Melting Pt:–22 °C
Density:1,4518 g/cm³ (20 °C)
Flash Pt:105°C
EINECS:206-233-6
UN:3265
ADR:8,III
:GHS05
CAS Number:312-94-7
Formula:C₈H₄ClF₃O
MDL Number:MFCD00000667