
3-Toluoyl chloride ≥98.0% (by GC, titration analysis)
TCIAT0309-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
:
:154.6 g/mol
:220°C
:–23 °C
:1,172 g/cm³ (20 °C)
:76°C
:Ambient
:216-976-8
:3265
:8,II
:GHS05
:1711-06-4
:C₈H₇ClO
:MFCD00000681