
2-Toluoyl chloride ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:933-88-0
:
Formula:C₈H₇ClO
MDL Number:MFCD00000668
:154.6 g/mol
Boiling Pt:213°C
Melting Pt:–18 °C
Density:1,186 g/cm³ (20 °C)
Flash Pt:101°C
EINECS:213-273-8
UN:3265
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together