Order Entry
Netherlands
ContactUsLinkComponent
4'-Methoxypropiophenone ≥97.0% (by GC)
Flammable
4'-Methoxypropiophenone ≥97.0% (by GC)
Catalog # TCIAM1719-25G
Supplier:  TCI
CAS Number:  
undefined
Flammable
4'-Methoxypropiophenone ≥97.0% (by GC)
Catalog # TCIAM1719-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White or Colorless to Yellow powder to lump to clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :164.2 g/mol
Boiling Pt:275°C
Melting Pt:∼37.5 °C
Density:~ 0,94 g/cm³ (20 °C)
Flash Pt:61°C
EINECS:204-512-7
UN:1325
ADR:4.1,III
 :GHS02
CAS Number:121-97-1
Formula:C₁₀H₁₂O₂
MDL Number:MFCD00009310