
1,3-Benzenedisulphonyl chloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:585-47-7
:
Formula:C₆H₄Cl₂O₄S₂
MDL Number:MFCD00041879
Boiling Pt:145°C
Melting Pt:63 °C
Storage Temperature:Refrigerator
EINECS:209-556-0
UN:3261
ADR:8,III
:275.13 g/mol
:GHS05
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:59.0 to 63.0 deg-C
- Solubility in Toluene:almost transparency
Specification Test Results
Frequently Bought Together