
3-(Dimethylamino)phenol ≥97.0% (by GC, titration analysis)
: TCI
:99-07-0
:
:C₈H₁₁NO
:MFCD00002264
:137.18 g/mol
:268°C
:65…72 °C
:Ambient
:202-727-0
:GHS07
- Appearance:White to Gray to Brown powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
- Melting point:84.0 to 87.0 deg-C
Frequently Bought Together