Order Entry
Netherlands
ContactUsLinkComponent
 
1,3-Dimethoxy-4-iodobenzene ≥98.0% (by GC)
Catalog # TCIAD0636-10G
Supplier:  TCI
CAS Number:  
undefined
1,3-Dimethoxy-4-iodobenzene ≥98.0% (by GC)
Catalog # TCIAD0636-10G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    39.0 to 42.0 deg-C
  • Solubility in Methanol:
    almost transparency

Specification Test Results

About this item

 :
 :264.06 g/mol
Boiling Pt:166°C
CAS Number:20469-63-0
Formula:C₈H₉IO₂
MDL Number:MFCD00059268