
Ethyl-3-iodobenzoate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:58313-23-8
:
Formula:C₉H₉IO₂
MDL Number:MFCD00044769
:276.07 g/mol
Boiling Pt:161°C
Density:1,66 g/cm³ (20 °C)
:GHS07
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together