
1-(Difluoromethoxy)-4-nitrobenzene
Catalog # TCIAD2601-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Light yellow to Brown to Dark green powder to lump
- Purity (GC):min. 98.0 %
- Freezing point:31.0 to 34.0 deg-C
Specification Test Results
About this item
:
:189.12 g/mol
:GHS07
CAS Number:1544-86-1
Formula:C₇H₅F₂NO₃
MDL Number:MFCD00085009