
3-Fluoro-4-methoxybenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:403-20-3
:
Formula:C₈H₇FO₃
MDL Number:MFCD00060675
:170.14 g/mol
Melting Pt:211.5 °C
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:210.0 to 224.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together