Order Entry
Netherlands
ContactUsLinkComponent
 
Ethyl-3-fluorobenzoate ≥98.0% (by GC)
  TCIAE0861-25G
 :  TCI
 :  
undefined
Ethyl-3-fluorobenzoate ≥98.0% (by GC)
  TCIAE0861-25G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

 

 

 :
 :168.17 g/mol
 :209°C
 :-34°C
 :–34°C
 :82°C
 :207-191-1
 :451-02-5
 :C₉H₉FO₂
 :MFCD00000336