Methyl-2-fluorobenzoate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:394-35-4
:
Formula:C₈H₇FO₂
MDL Number:MFCD00017913
:154.14 g/mol
Boiling Pt:102°C
Flash Pt:94°C
EINECS:206-894-0
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together