
4-Hydroxybenzhydrazide ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:5351-23-5
:
Formula:C₇H₈N₂O₂
MDL Number:MFCD00007605
:152.15 g/mol
Boiling Pt:370 °C (1013 hPa)
Melting Pt:268°C
Storage Temperature:Ambient
EINECS:226-326-5
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light red powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 96.0 %
Specification Test Results
Frequently Bought Together