
3-Nitrobenzhydrazide ≥98.0% (by titrimetric analysis)
Catalog # TCIAN0147-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (Bromination Method):min. 98.0 %
- Melting point:153.0 to 157.0 deg-C
Specification Test Results
About this item
:
:181.15 g/mol
EINECS:210-572-5
:GHS07
CAS Number:618-94-0
Formula:C₇H₇N₃O₃
MDL Number:MFCD00017059