


3,5-Dinitrobenzoyl chloride ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:99-33-2
:
Formula:C₇H₃ClN₂O₅
MDL Number:MFCD00007248
:230.56 g/mol
Merck Index:14,03277
Boiling Pt:339 °C (1013 hPa)
Melting Pt:68…70 °C
Density:1,33 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-750-6
UN:3261
ADR:8,III
:GHS05
:GHS07
:GHS08
Specification Test Results
- Appearance:White to Orange to Green powder to lump
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:68.0 to 71.0 deg-C
Specification Test Results
Frequently Bought Together