
3-Acetamidobenzoic acid ≥99.0% (by HPLC, titration analysis)
TCIAA0012-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 99.0 area%
- Purity (Neutralization titration):min. 99.0 %
- Melting point:245.0 to 251.0 deg-C
:
:179.18 g/mol
:209-600-9
:GHS07
:587-48-4
:C₉H₉NO₃
:MFCD00013983