

3-Cyanobenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:56542-67-7
:
Formula:C₇H₄ClNO₂S
MDL Number:MFCD00203480
Boiling Pt:111°C
UN:2928
ADR:6.1,II
:201.63 g/mol
:GHS05
:GHS06
Specification Test Results
- Appearance:White to Light yellow to Dark green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:48.0 to 53.0 deg-C
Specification Test Results
Frequently Bought Together