
Methyl-4-cyanobenzoate ≥98.0% (by GC)
: TCI
:1129-35-7
:
:C₉H₇NO₂
:MFCD00001823
:161.16 g/mol
:142°C
:67 °C
:214-443-4
:6.1,III
:GHS07
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:65.0 to 69.0 deg-C
Frequently Bought Together