Order Entry
Netherlands
ContactUsLinkComponent
3,5-Dichlorobenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
Corrosive
3,5-Dichlorobenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Corrosive
3,5-Dichlorobenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to lump
  • Purity (GC):
    min. 98.0 %
  • Purity (Argentmetric Titration):
    min. 98.0 %

Specification Test Results

Frequently Bought Together