
2,5-Dichlorobenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Certificaten
Over dit artikel
CAS-nummer:5402-73-3
:
Formule:C₆H₃Cl₃O₂S
MDL-nummer:MFCD00007429
:245.51 g/mol
Smelten Pt:39 °C
Temperatuur bij opslag:Refrigerator
EINECS:226-453-6
UN:3261
ADR:8,III
:GHS05
Specificatie testresultaten
- Appearance:White or Colorless to Light yellow powder to lump to clear liquid
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:35.0 to 39.0 deg-C
- Solubility in Toluene:almost transparency
Specificatie testresultaten
Frequently Bought Together