
2,3-Dichlorobenzenesulphonyl chloride ≥96.0% (by GC, titration analysis)
Catalog # TCIAD3568-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to lump
- Purity (GC):min. 96.0 %
- Purity (Argentmetric Titration):min. 96.0 %
- Melting point:60.0 to 64.0 deg-C
Specification Test Results
About this item
:
:245.51 g/mol
Boiling Pt:140 - 142 °C (2 -rr)
Melting Pt:61…63 °C
Storage Temperature:Refrigerator
UN:3261
ADR:8,II
:GHS05
CAS Number:82417-45-6
Formula:C₆H₃Cl₃O₂S
MDL Number:MFCD00051844