
2-Chlorobenzhydrazide ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:5814-05-1
:
Formula:C₇H₇ClN₂O
MDL Number:MFCD00007597
:170.6 g/mol
EINECS:227-380-2
:GHS07
Specification Test Results
- Appearance:White to Gray to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:116.0 to 120.0 deg-C
Specification Test Results
Frequently Bought Together