Order Entry
Netherlands
ContactUsLinkComponent
 
Methyl-4-bromobenzoate ≥98.0% (by GC)
 :  TCI
 
undefined
Methyl-4-bromobenzoate ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    79.0 to 81.0 deg-C
  • Solubility in Methanol:
    almost transparency

 

Frequently Bought Together