Methyl-2-bromobenzoate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:610-94-6
:
Formula:C₈H₇BrO₂
MDL Number:MFCD00016328
Boiling Pt:253°C
Density:1,51 g/cm³ (20 °C)
:215.05 g/mol
EINECS:210-241-5
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together