Order Entry
Netherlands
ContactUsLinkComponent
Ethyl-2-bromobenzoate ≥98.0% (by GC)
Ethyl-2-bromobenzoate ≥98.0% (by GC)
  TCIAB1514-25G
 :  TCI
 :  
Ethyl-2-bromobenzoate ≥98.0% (by GC)
  TCIAB1514-25G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 98.0 %

 

 

 :
 :229.07 g/mol
 :255°C
 :1,441 g/cm³ (20 °C)
 :228-034-3
 :6091-64-1
 :C₉H₉BrO₂
 :MFCD00015443