Order Entry
Netherlands
ContactUsLinkComponent
 
 
 
 
2-Hexyn-1-ol ≥97.0% (by GC)
  TCIAH0465-5ML
 :  TCI
 :  
undefined
 
 
 
2-Hexyn-1-ol ≥97.0% (by GC)
  TCIAH0465-5ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

 

 

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 97.0 %

 

 

 :
 :98.14 g/mol
 :167°C
 :52°C
 :212-128-6
 :1987
 :3,III
 :GHS02
 :GHS05
 :GHS07
 :764-60-3
 :C₆H₁₀O
 :MFCD00039540