Order Entry
Netherlands
ContactUsLinkComponent
 
cis-4-Hexen-1-ol ≥95.0% (by GC)
Catalog # TCIAH0754-10ML
Supplier:  TCI
CAS Number:  
undefined
cis-4-Hexen-1-ol ≥95.0% (by GC)
Catalog # TCIAH0754-10ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • Seq:
    2275

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

About this item

 :
Boiling Pt:158°C
Flash Pt:65°C
ADR:3,III
 :100.16 g/mol
EINECS:213-187-0
CAS Number:928-91-6
Formula:C₆H₁₂O
MDL Number:MFCD00148974