Order Entry
Netherlands
ContactUsLinkComponent
cis-3-Octen-1-ol ≥95.0% (by GC)
cis-3-Octen-1-ol ≥95.0% (by GC)
Catalog # TCIAO0248-25ML
Supplier:  TCI
CAS Number:  
undefined
cis-3-Octen-1-ol ≥95.0% (by GC)
Catalog # TCIAO0248-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

About this item

 :
 :128.21 g/mol
Boiling Pt:89°C
Flash Pt:82°C
Storage Temperature:Refrigerator
EINECS:243-533-6
CAS Number:20125-84-2
Formula:C₈H₁₆O
MDL Number:MFCD00014062