

2,2-Bis(bromomethyl)-1,3-propanediol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:3296-90-0
:
Formula:C₅H₁₀Br₂O₂
MDL Number:MFCD00004688
:261.94 g/mol
Boiling Pt:371 °C (1013 hPa)
Melting Pt:107…109 °C
Density:2,2 g/cm³ (20 °C)
EINECS:221-967-7
:GHS07
:GHS08
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:109.0 to 113.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together