
1,3-Diethoxy-2-propanol ≥98.0% (by GC)
TCIAD0460-10ML
: TCI
:
- Pack type:Glass bottle
- Pk:10 mL
- Seq:2275
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
:
:191°C
:0,95 g/cm³ (20 °C)
:58°C
:3,III
:148.2 g/mol
:223-736-6
:1993
:GHS02
:4043-59-8
:C₇H₁₆O₃
:MFCD00026934