


1-(Dimethylamino)-2-propanol ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:108-16-7
:
Formula:C₅H₁₃NO
MDL Number:MFCD00004532
:103.16 g/mol
Boiling Pt:125°C
Melting Pt:–85 °C
Density:0,85 g/cm³ (20 °C)
Flash Pt:32°C
Storage Temperature:Ambient
EINECS:203-556-4
UN:2734
ADR:8,II
:GHS02
:GHS05
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together