Order Entry
Netherlands
ContactUsLinkComponent
1-(Diethylamino)-2-propanol ≥98.0% (by GC, titration analysis)
Flammable
Corrosive
1-(Diethylamino)-2-propanol ≥98.0% (by GC, titration analysis)
Catalog # TCIAD0471-25ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Corrosive
1-(Diethylamino)-2-propanol ≥98.0% (by GC, titration analysis)
Catalog # TCIAD0471-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :131.22 g/mol
Boiling Pt:159°C
Melting Pt:13.5 °C
Density:0,8677 g/cm³ (0 °C)
Flash Pt:33°C
EINECS:224-537-7
UN:2734
ADR:8,II
 :GHS02
 :GHS05
CAS Number:4402-32-8
Formula:C₇H₁₇NO
MDL Number:MFCD00004535