

Ethyl trifluoromethanesulphonate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:425-75-2
:
Formula:C₃H₅F₃O₃S
MDL Number:MFCD00000410
:178.13 g/mol
Boiling Pt:115°C
Density:1,374 g/cm³ (0 °C)
Flash Pt:35°C
EINECS:207-037-3
UN:2924
ADR:3,III
:GHS02
:GHS05
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together