
Triethylsilyl trifluoromethanesulphonate ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:79271-56-0
:
Formula:C₇H₁₅F₃O₃SSi
MDL Number:MFCD00000407
Boiling Pt:79°C
Density:1,225 g/cm³ (20 °C)
Flash Pt:72°C
EINECS:279-124-4
UN:1760
ADR:8,III
:264.34 g/mol
:GHS05
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together