D-Methionine ≥99.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:348-67-4
:
Formula:C₅H₁₁NO₂S
MDL Number:MFCD00002622
:149.21 g/mol
Merck Index:13,06004
Boiling Pt:278 °C (1013 hPa)
Melting Pt:136 °C
EINECS:206-483-6
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 99.0 %
- Specific rotation [a]20/D:-23.0 to -25.0 deg (C=4,HCl (1+1))
- Solubility in hot Water:almost transparency
Specification Test Results
Related Information
Frequently Bought Together