
Pyruvic acid ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:127-17-3
:
Formula:C₃H₄O₃
MDL Number:MFCD00002585
:88.06 g/mol
Merck Index:14,08021
Boiling Pt:62°C
Melting Pt:13°C
Density:1,58 g/cm³ (20 °C)
Flash Pt:82°C
Storage Temperature:Refrigerator
EINECS:204-824-3
UN:3265
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Colorless to Yellow to Green clear liquid
- Purity (GC):min. 97.0 %
- Purity (Iodometric Titration):min. 95.0 %
Specification Test Results
Frequently Bought Together