
Methyl pyruvate ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:600-22-6
:
Formula:C₄H₆O₃
MDL Number:MFCD00008754
:102.09 g/mol
Boiling Pt:138°C
Density:1,154 g/cm³ (0 °C)
Flash Pt:39°C
Storage Temperature:Refrigerator
EINECS:209-987-4
UN:3272
ADR:3,III
:GHS02
Specification Test Results
- Appearance:Colorless to Orange to Green clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together