(S,S)-(+)-2,5-Hexanediol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:34338-96-0
:
Formula:C₆H₁₄O₂
MDL Number:MFCD00082583
:118.18 g/mol
Boiling Pt:71°C
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:52.0 to 56.0 deg-C
- Specific rotation [a]20/D:+23.0 to +27.0 deg (C=1, ethanol)
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together