(R,R)-(-)-2,5-Hexanediol ≥98.0% (by GC)
Catalog # TCIAH1032-1G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White or Colorless powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:52.0 to 56.0 deg-C
- Specific rotation [a]20/D:-23.0 to -26.0 deg (C=1, EtOH)
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
:118.18 g/mol
Storage Temperature:Refrigerator
CAS Number:17299-07-9
Formula:C₆H₁₄O₂
MDL Number:MFCD00142339