N-Acetyl-L-cysteine ≥98.0% (by HPLC, titration analysis)
: TCI
:616-91-1
:
:C₅H₉NO₃S
:MFCD00004880
:163.2 g/mol
:14,00088
:371 °C (1013 hPa)
:106…108 °C
:1,48 g/cm³ (20 °C)
:Ambient
:210-498-3
- Appearance:White to Almost white powder to crystaline
- Purity (HPLC):min. 98.0 area%
- Purity (Iodometric Titration):min. 98.0 %
- Melting point:108.0 to 113.0 deg-C
- Specific rotation [a]20/D:+21.0 to +27.0 deg (C=5, JPC Method)
Frequently Bought Together