
2,3,5-Triphenyltetrazolium chloride (TTC) ≥98.0% (by titrimetric analysis)
: TCI
:298-96-4
:
:C₁₉H₁₅ClN₄
:MFCD00011963
:14,09744
:? 360 °C (1013 hPa)
:235 °C
:334.81 g/mol
:Refrigerator
:206-071-6
:GHS07
- Appearance:White to Orange to Green powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 % (calcd.on dried substance)
- Absorbance:min. 0.700 (10 mg/L, 243.0 to 247.0 nm) (calcd.on dried substance)
- Drying loss:max. 5.0 %
- Sensitiveness:to pass test
Frequently Bought Together