

(±)-α-Methylbenzylamine ≥98.0% (by GC, titration analysis)
: TCI
:618-36-0
:
:C₆H₅CH(NH₂)CH₃
:MFCD00008069
:121.18 g/mol
:14,06026
:185°C
:–65 °C
:0,95 g/cm³ (20 °C)
:69°C
:Ambient
:210-545-8
:2922
:8,II
:GHS05
:GHS07
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Frequently Bought Together