(±)-1,2-Dodecanediol ≥93.0% (by GC)
: TCI
:1119-87-5
:
:C₁₂H₂₆O₂
:MFCD00004726
:202.34 g/mol
:214-289-8
- Appearance:White to Almost white powder to crystaline
- Purity (GC):min. 93.0 %
- Melting point:57.0 to 61.0 deg-C
Frequently Bought Together