DL-Methionine ≥99.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:59-51-8
:
Formula:C₅H₁₁NO₂S
MDL Number:MFCD00063096
:149.21 g/mol
Merck Index:14,05975
Melting Pt:∼136 °C
Density:1,34 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-432-1
Specification Test Results
- Appearance:White powder to crystal
- Purity (HPLC):min. 99.0 area%
- Purity (Nonaqueous Titration):min. 99.0 %
- Solubility in hot Water:almost transparency
Specification Test Results
Frequently Bought Together