

Triphenylphosphine ≥95.0% (by titrimetric analysis)
: TCI
:603-35-0
:
:C₁₈H₁₅P
:MFCD00003043
:262.29 g/mol
:14,09743
:260°C
:79 °C
:1,194 g/cm³ (20 °C)
:180 °C
:Ambient
:210-036-0
:GHS07
:GHS08
- Appearance:White powder to crystal
- Purity (Iodometric Titration):min. 95.0 %
- Melting point:80.0 to 82.0 deg-C
- Solubility in Toluene:very faint turbidity
Frequently Bought Together