
4-Chloro-2-fluorocinnamic acid ≥97.0% (by GC, titration analysis)
Catalog # TCIAC2200-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Neutralization titration):min. 97.0 %
- Melting point:213.0 to 217.0 deg-C
Specification Test Results
About this item
:
:200.6 g/mol
:GHS07
CAS Number:202982-65-8
Formula:C₉H₆ClFO₂
MDL Number:MFCD00143287