Order Entry
Netherlands
ContactUsLinkComponent
 
Irritant
3,4,5-Trimethoxycinnamic acid ≥98.0% (by GC, titration analysis)
Catalog # TCIAT1104-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
3,4,5-Trimethoxycinnamic acid ≥98.0% (by GC, titration analysis)
Catalog # TCIAT1104-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    127.0 to 130.0 deg-C

Specification Test Results

About this item

 :
 :238.24 g/mol
Boiling Pt:367 °C (1013 hPa)
Melting Pt:127 °C
EINECS:201-999-8
 :GHS07
CAS Number:90-50-6
Formula:C₁₂H₁₄O₅
MDL Number:MFCD00004388